Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

E0986-5G E0986-25G
TCIAE0986-5GEA 16 EUR
TCIAE0986-5G TCIAE0986-25G
Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate ≥98.0% (by GC, titration analysis)
Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate
Formula: C₇H₁₁N₃O₂
Número MDL: MFCD00051652
Núm. CAS: 31037-02-2

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 99.0 to 103.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR