(R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-4-hydroxybenzeneacetic acid ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
(R)-2-(4-Ethyl-2,3-dioxopiperazine-1-carboxamido)-2-(4-hydroxyphenyl)acetic acid, (R)-(-)-2-(4-Ethyl-2,3-dioxopiperazine-1-carboxamido)-2-(4-hydroxyphenyl)acetic acid
E0994-5G
E0994-1G
TCIAE0994-5GEA
72
EUR
TCIAE0994-5G
TCIAE0994-1G
(R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-4-hydroxybenzeneacetic acid ≥98.0% (by HPLC, titration analysis)
(R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-4-hydroxybenzeneacetic acid
Formula:
C₁₅H₁₇N₃O₆ MW: 335,32 g/mol |
Número MDL:
MFCD00792472 Núm. CAS: 62893-24-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | -84.0 to -87.0 deg (C=1,MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...