(S,S)-N,N'-Bis(1-hydroxy-2-butyl)ethylenediamine dihydrochloride ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Ethambutol Hydrochloride, (2S,2'S)-2,2'-(Ethane-1,2-diylbis(azanediyl))bis(butan-1-ol) dihydrochloride, (S,S)-2,2'-(1,2-Ethanediyldiimino)bis-1-butanol Dihydrochloride

E1011-5G E1011-25G
TCIAE1011-5GEA 26 EUR
TCIAE1011-5G TCIAE1011-25G
(S,S)-N,N'-Bis(1-hydroxy-2-butyl)ethylenediamine dihydrochloride ≥98.0% (by titrimetric analysis)
(S,S)-N,N'-Bis(1-hydroxy-2-butyl)ethylenediamine dihydrochloride
Formula: C₁₀H₂₄N₂O₂·₂HCl
MW: 277,23 g/mol
Número MDL: MFCD00216025
Núm. CAS: 1070-11-7
EINECS: 213-970-7
Índice Merck: 14,03720

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 199.0 to 204.0 deg-C
Specific rotation [a]20/D +5.0 to +7.0 deg (C=10, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR