Erdosteine ≥98.0% (by GC, titration analysis)

Proveedor: TCI

Sinónimos: N-(Tetrahydro-2-oxo-3-thienyl)-3-thiapentanedioic Acid Monoamide, 2-((2-Oxo-2-((2-oxotetrahydrothiophen-3-yl)amino)ethyl)thio)acetic acid

E1026-1G E1026-5G
TCIAE1026-1GEA 50 EUR
TCIAE1026-1G TCIAE1026-5G
Erdosteine ≥98.0% (by GC, titration analysis)
Erdosteine
Formula: C₈H₁₁NO₄S₂
MW: 249,30 g/mol
Número MDL: MFCD00867664
Núm. CAS: 84611-23-4
Índice Merck: 14,03647

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 160.0 to 164.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR