Ethyl-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
Ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate, Ethyl 5-Formyl-2,4-dimethyl-3-pyrrolecarboxylate, Ethyl 5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate, 5-Formyl-2,4-dimethyl-3-pyrrolecarboxylic acid ethyl ester
E1051-1G
E1051-5G
TCIAE1051-1GEA
39
EUR
TCIAE1051-1G
TCIAE1051-5G
Ethyl-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate ≥98.0% (por GC)
Ethyl-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate
Formula:
C₁₀H₁₃NO₃ MW: 195,22 g/mol |
Número MDL:
MFCD00030352 Núm. CAS: 2199-59-9 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 164.0 to 168.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...