O-Ethyl-N,N'-diisopropylisourea ≥98.0% (by GC, titration analysis)
Proveedor: TCI
E1058-5ML
E1058-25ML
TCIAE1058-5MLEA
67
EUR
TCIAE1058-5ML
TCIAE1058-25ML
O-Ethyl-N,N'-diisopropylisourea ≥98.0% (by GC, titration analysis)
O-Ethyl-N,N'-diisopropylisourea
Formula:
C₉H₂₀N₂O Pto. Ebullición: 65°C Temperatura de almacenaje: Nevera |
Número MDL:
MFCD24554718 Núm. CAS: 60683-30-9 |
Resultados test especificaciones
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...