Ethylboronic acid pinacol ester ≥95.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-Ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, (4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)ethane, Ethylboronic acid Pinacol ester
E1116-1G
E1116-5G
TCIAE1116-1GEA
38
EUR
TCIAE1116-1G
TCIAE1116-5G
Ethylboronic acid pinacol ester ≥95.0% (by GC, titration analysis)
Ethylboronic acid pinacol ester
Formula:
C₈H₁₇BO₂ MW: 156,03 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD19441153 Núm. CAS: 82954-89-0 UN: 3272 ADR: 3,III |
Resultados test especificaciones
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...