PDE5-IN-7 ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 5-(2-Ethoxyphenyl)-1-methyl-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-7(6H)-one, 5-(2-Ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]-7-pyrimidinone

E1134-5G E1134-25G
TCIAE1134-5GEA 41 EUR
TCIAE1134-5G TCIAE1134-25G
PDE5-IN-7 ≥98.0% (by HPLC, titration analysis)
PDE5-IN-7
Formula: C₁₇H₂₀N₄O₂
MW: 312,37 g/mol
Número MDL: MFCD04088989
Núm. CAS: 139756-21-1

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 143.0 to 147.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR