L-Glutamic acid 5-ethyl ester ≥97.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
5-Ethyl L-Glutamate, L-Glutamic acid 5-Ethyl ester, H-Glu(OEt)-OH
E1155-5G
E1155-25G
TCIAE1155-5GEA
22
EUR
TCIAE1155-5G
TCIAE1155-25G
L-Glutamic acid 5-ethyl ester ≥97.0% (by titrimetric analysis)
L-Glutamic acid 5-ethyl ester
Formula:
C₇H₁₃NO₄ MW: 175,2 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00037217 Núm. CAS: 1119-33-1 EINECS: 214-274-6 Índice Merck: 14,04470 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 97.0 % |
Specific rotation [a]20/D | +11.0 to +14.0 deg (C=1,H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...