trans,trans-4-(4-Ethoxy-2,3-difluorophenyl)-4'-n-propylbicyclohexyl ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
(trans,trans)-4-(4-Ethoxy-2,3-difluorophenyl)-4'-propyl-1,1'-bi(cyclohexane), trans,trans-4-(4-Ethoxy-2,3-difluorophenyl)-4'-propylbicyclohexyl
E1157-1G
E1157-5G
TCIAE1157-1GEA
48
EUR
TCIAE1157-1G
TCIAE1157-5G
trans,trans-4-(4-Ethoxy-2,3-difluorophenyl)-4'-n-propylbicyclohexyl ≥98.0% (por GC)
trans,trans-4-(4-Ethoxy-2,3-difluorophenyl)-4'-n-propylbicyclohexyl
Formula:
C₂₃H₃₄F₂O MW: 364,521 g/mol |
Número MDL:
MFCD18433524 Núm. CAS: 123560-48-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 77.0 to 81.0 deg-C |
Mesomorphic range | 78.0 to 84.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...