Ethyl-2-(2-aminothiazol-4-yl)-2-oxoacetate ≥96.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2-(2-Amino-4-thiazolyl)-2-oxoacetic acid ethyl ester, Ethyl 2-(2-Amino-4-thiazolyl)-2-oxoacetate, Ethyl 2-(2-aminothiazol-4-yl)-2-oxoacetate

E1159-5G E1159-25G
TCIAE1159-5GEA 93 EUR
TCIAE1159-5G TCIAE1159-25G
Ethyl-2-(2-aminothiazol-4-yl)-2-oxoacetate ≥96.0% (by HPLC, titration analysis)
Ethyl-2-(2-aminothiazol-4-yl)-2-oxoacetate
Formula: C₇H₉NO₄S
MW: 200,2150 g/mol
Número MDL: MFCD00010414
Núm. CAS: 64987-08-2

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 144.0 to 148.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR