1-Ethynyl-3,5-dimethoxybenzene ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
3,5-Dimethoxyphenylacetylene
E1175-200MG
E1175-1G
TCIAE1175-200MGEA
36
EUR
TCIAE1175-200MG
TCIAE1175-1G
1-Ethynyl-3,5-dimethoxybenzene ≥98.0% (por GC)
1-Ethynyl-3,5-dimethoxybenzene
Formula:
C₁₀H₁₀O₂ MW: 162,19 g/mol |
Número MDL:
MFCD03839985 Núm. CAS: 171290-52-1 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 44.0 to 48.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...