2-Ethoxy-5-methylbenzeneboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-Ethoxy-5-methylbenzeneboronic acid, 2-Ethoxy-5-methylphenylboronic acid
E1189-1G
E1189-5G
TCIAE1189-1GEA
50
EUR
TCIAE1189-1G
TCIAE1189-5G
2-Ethoxy-5-methylbenzeneboronic acid (contains varying amounts of Anhydride)
2-Ethoxy-5-methylbenzeneboronic acid
Formula:
C₉H₁₃BO₃ MW: 180,01 g/mol |
Número MDL:
MFCD00196832 Núm. CAS: 123291-97-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 111.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...