4-Ethoxy-2,3-difluorophenol ≥98.0% (by GC, titration analysis)
Proveedor: TCI
E1193-1G
E1193-5G
TCIAE1193-1GEA
26
EUR
TCIAE1193-1G
TCIAE1193-5G
4-Ethoxy-2,3-difluorophenol ≥98.0% (by GC, titration analysis)
4-Ethoxy-2,3-difluorophenol
Formula:
C₈H₈F₂O₂ MW: 174,14 g/mol |
Número MDL:
MFCD07368025 Núm. CAS: 126163-56-2 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 76.0 to 80.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...