Ethyl alpha-Iodophenylacetate ≥97.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: 2-Iodo-2-phenylacetic acid Ethyl ester, alpha-Iodophenylacetic acid Ethyl ester, Ethyl 2-Iodo-2-phenylacetate

E1200-5G E1200-25G
TCIAE1200-5GEA 49 EUR
TCIAE1200-5G TCIAE1200-25G
Ethyl alpha-Iodophenylacetate ≥97.0% (by HPLC, titration analysis)
Ethyl alpha-Iodophenylacetate
Formula: C₁₀H₁₁IO₂
MW: 290,10 g/mol
Temperatura de almacenaje: Congelador
Núm. CAS: 78489-65-3
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff471cd7036f62e93a65dfbee84fb30ec90ccd2&vid=8ff471cd7036f62e93a65dfbee84fb30ec90ccd2&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown clear liquid
Purity (HPLC) min. 97.0 area%
Purity (Argentmetric Titration) min. 97.0 %
Specific gravity (20/20) 1.5630 to 1.5680
Refractive index n20/D 1.5780 to 1.5820

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR