3-Ethoxy-5-fluorobenzeneboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
(3-Ethoxy-5-fluorophenyl)boronic acid, 3-Ethoxy-5-fluorophenylboronic acid, 3-Ethoxy-5-fluorobenzeneboronic acid
E1205-200MG
E1205-1G
TCIAE1205-200MGEA
40
EUR
TCIAE1205-200MG
TCIAE1205-1G
3-Ethoxy-5-fluorobenzeneboronic acid (contains varying amounts of Anhydride)
3-Ethoxy-5-fluorobenzeneboronic acid
Formula:
C₈H₁₀BFO₃ MW: 183,97 g/mol |
Número MDL:
MFCD07363779 Núm. CAS: 850589-53-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 111.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...