1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid, 1-(Ethoxycarbonyl)piperidine-4-carboxylic acid

E1208-1G E1208-5G
TCIAE1208-1GEA 27 EUR
TCIAE1208-1G TCIAE1208-5G
1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid ≥98.0% (by GC, titration analysis)
1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid
Formula: C₉H₁₅NO₄
MW: 201,22 g/mol
Número MDL: MFCD07339354
Núm. CAS: 118133-15-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce94bbffa456f39138755102d291a71694ceb5d6&vid=ce94bbffa456f39138755102d291a71694ceb5d6&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 67.0 to 71.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR