Ácido 2,6-difluoro-3-etoxifenilborónico (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
3-Ethoxy-2,6-difluorophenylboronic acid, 3-Ethoxy-2,6-difluorobenzeneboronic acid
E1210-1G
E1210-5G
TCIAE1210-1GEA
34
EUR
TCIAE1210-1G
TCIAE1210-5G
Ácido 2,6-difluoro-3-etoxifenilborónico (contains varying amounts of Anhydride)
Ácido 2,6-difluoro-3-etoxifenilborónico
Formula:
C₈H₉BF₂O₃ MW: 201,96 g/mol |
Número MDL:
MFCD07369735 Núm. CAS: 849062-00-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...