Ethyl-3-(pyridin-2-ylamino)propanoate ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: 3-(2-Pyridylamino)propionic acid ethyl ester, Ethyl 3-(2-Pyridylamino)propionate, Ethyl 3-(pyridin-2-ylamino)propanoate

E1214-5G E1214-25G
TCIAE1214-5GEA 23 EUR
TCIAE1214-5G TCIAE1214-25G
Ethyl-3-(pyridin-2-ylamino)propanoate ≥98.0% (by titrimetric analysis)
Ethyl-3-(pyridin-2-ylamino)propanoate
Formula: C₁₀H₁₄N₂O₂
MW: 194,2300 g/mol
Pto. Ebullición: 125°C
Número MDL: MFCD09833621
Núm. CAS: 103041-38-9

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to lump
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 51.0 to 55.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR