3,5-Dimethyl-4-ethoxyphenylboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: 4-Ethoxy-3,5-dimethylbenzeneboronic acid, 4-Ethoxy-3,5-dimethylphenylboronic acid, (4-Ethoxy-3,5-dimethylphenyl)boronic acid, 3,5-Dimethyl-4-ethoxybenzeneboronic acid

E1216-1G E1216-5G
TCIAE1216-1GEA 27 EUR
TCIAE1216-1G TCIAE1216-5G
3,5-Dimethyl-4-ethoxyphenylboronic acid (contains varying amounts of Anhydride)
3,5-Dimethyl-4-ethoxyphenylboronic acid
Formula: C₁₀H₁₅BO₃
MW: 194,04 g/mol
Número MDL: MFCD02179463
Núm. CAS: 850568-59-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf493fdf4b225c6c05a758043f4d1ffe0f21f049&vid=cf493fdf4b225c6c05a758043f4d1ffe0f21f049&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR