Diioduro de etilendiamonio ≥98.0% (by total nitrogen and titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Ethylenediamine dihydroiodide
E1222-1G
E1222-5G
TCIAE1222-1GEA
32
EUR
TCIAE1222-1G
TCIAE1222-5G
Diioduro de etilendiamonio ≥98.0% (by total nitrogen and titration analysis)
Diioduro de etilendiamonio
Formula:
C₂H₁₀I₂N₂ MW: 315,92 g/mol |
Número MDL:
MFCD00035535 Núm. CAS: 5700-49-2 EINECS: 227-186-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...