Ethyl (3-Hydroxyphenyl)carbamate ≥97.0% (por GC)
Proveedor: TCI
Sinónimos:
(3-Hydroxyphenyl)carbamic acid Ethyl ester
E1227-1G
E1227-5G
TCIAE1227-1GEA
102
EUR
TCIAE1227-1G
TCIAE1227-5G
Ethyl (3-Hydroxyphenyl)carbamate ≥97.0% (por GC)
Ethyl (3-Hydroxyphenyl)carbamate
Formula:
C₉H₁₁NO₃ MW: 181,19 g/mol |
Número MDL:
MFCD00463773 Núm. CAS: 7159-96-8 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 93.0 to 97.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...