Ácido (5-etoxi-2-fluorofenil)metabórico ≥98.0% (by HPLC, titration analysis) (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: Dihidroxi-5-etoxi-2-fluorofenilborano

E1235-1G E1235-5G
TCIAE1235-1GEA 41 EUR
TCIAE1235-1G TCIAE1235-5G
Ácido (5-etoxi-2-fluorofenil)metabórico ≥98.0% (by HPLC, titration analysis) (contains varying amounts of Anhydride)
Ácido (5-etoxi-2-fluorofenil)metabórico
Formula: C₈H₁₀BFO₃
MW: 183,97 g/mol
Número MDL: MFCD05664321
Núm. CAS: 900174-60-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea998aa46a69e0165ca8e45ba9fe23f6167c1b5&vid=bea998aa46a69e0165ca8e45ba9fe23f6167c1b5&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) 98.0 to 111.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR