1-Ethyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2-(1-Ethyl-1H-pyrazol-4yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Ethyl-1H-pyrazole-4-boronic acid Pinacol ester, 1-Ethyl-1H-pyrazole-4-boronic acid, pinacol ester, 1-Ethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

E1348-1G E1348-5G
TCIAE1348-1GEA 42 EUR
TCIAE1348-1G TCIAE1348-5G
1-Ethyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
1-Ethyl-1H-pyrazole-4-boronic acid pinacol ester
Formula: C₁₁H₁₉BN₂O₂
MW: 222,0918 g/mol
Número MDL: MFCD09864188
Núm. CAS: 847818-70-6

Order Now

Resultados test especificaciones

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR