Ethyl 2-[trans-4-[(tert-Butoxycarbonyl)amino]cyclohexyl]acetate ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: Ethyl 2-[trans-4-(Boc-amino)cyclohexyl]acetate, Ethyl 2-(trans-4-((tert-butoxycarbonyl)amino)cyclohexyl)acetate, 2-[trans-4-(Boc-amino)cyclohexyl]acetic acid Ethyl ester, 2-[trans-4-[(tert-Butoxycarbonyl)amino]cyclohexyl]acetic acid Ethyl ester

E1387-200MG E1387-1G
TCIAE1387-200MGEA 56 EUR
TCIAE1387-200MG TCIAE1387-1G
Ethyl 2-[trans-4-[(tert-Butoxycarbonyl)amino]cyclohexyl]acetate ≥98.0% (por GC)
Ethyl 2-[trans-4-[(tert-Butoxycarbonyl)amino]cyclohexyl]acetate
Formula: C₁₅H₂₇NO₄
Número MDL: MFCD20491405
Núm. CAS: 946598-34-1

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 73.0 to 77.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR