Ethyl 2-(3-Cyano-4-isobutoxyphenyl)-4-methyl-5-thiazolecarboxylate ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

Sinónimos: 2-(3-Cyano-4-isobutoxyphenyl)-4-methyl-5-thiazolecarboxylic acid Ethyl ester, Ethyl 2-(3-cyano-4-isobutoxyphenyl)-4-methyl-5-thiazolecarboxylate

E1414-25G E1414-5G
TCIAE1414-25GEA 179 EUR
TCIAE1414-25G TCIAE1414-5G
Ethyl 2-(3-Cyano-4-isobutoxyphenyl)-4-methyl-5-thiazolecarboxylate ≥98.0% (by HPLC, total nitrogen)
Ethyl 2-(3-Cyano-4-isobutoxyphenyl)-4-methyl-5-thiazolecarboxylate
Formula: C₁₈H₂₀N₂O₃S
MW: 344,43 g/mol
Número MDL: MFCD09833701
Núm. CAS: 160844-75-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 174.0 to 178.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR