9-Fluorenone oxime ≥97.0% (by GC, total nitrogen)

Proveedor: TCI

Sinónimos: 9H-Fluoren-9-one oxime

F0023-1G F0023-5G
TCIAF0023-1GEA 16 EUR
TCIAF0023-1G TCIAF0023-5G
9-Fluorenone oxime ≥97.0% (by GC, total nitrogen)
9-Fluorenone oxime
Formula: C₁₃H₉NO
MW: 195,22 g/mol
Número MDL: MFCD00016356
Núm. CAS: 2157-52-0
EINECS: 218-471-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31e3e05905a80184bdb5d09107ad0a0b82fb10eb&vid=31e3e05905a80184bdb5d09107ad0a0b82fb10eb&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (with Total Nitrogen) min. 97.0 %
Melting point 190.0 to 197.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR