Fluorescin ≥95.0% (por GC) reagent for oxidases and peroxidases

Proveedor: TCI
Atención

F0028-25G F0028-5G
TCIAF0028-25GEA 416 EUR
TCIAF0028-25G TCIAF0028-5G
Fluorescin ≥95.0% (por GC) reagent for oxidases and peroxidases
Fluorescin
Formula: C₂₀H₁₄O₅
Número MDL: MFCD00046931
Núm. CAS: 518-44-5
EINECS: 208-252-5
Índice Merck: 14,04160

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR