Formamidine hydrochloride ≥97.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Formimidamide hydrochloride

F0103-25G F0103-5G
TCIAF0103-25GEA 114 EUR
TCIAF0103-25G TCIAF0103-5G
Formamidine hydrochloride ≥97.0% (by titrimetric analysis)
Formamidine hydrochloride
Formula: CH₄N₂
MW: 80,52 g/mol
Número MDL: MFCD00012865
Núm. CAS: 6313-33-3
EINECS: 228-639-2

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 97.0 %
Melting point 83.0 to 88.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR