N-Formyl-DL-phenylalanine ≥99.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
2-Formamido-3-phenylpropanoic acid
F0126-25G
F0126-5G
TCIAF0126-25GEA
149
EUR
TCIAF0126-25G
TCIAF0126-5G
N-Formyl-DL-phenylalanine ≥99.0% (by titrimetric analysis)
N-Formyl-DL-phenylalanine
Formula:
C₁₀H₁₁NO₃ MW: 193.20 g/mol |
Número MDL:
MFCD00063281 Núm. CAS: 4289-95-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 167.0 to 171.0 deg-C |
Solubility in hot EtOH | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...