2-Fluoro-β-alanine hydrochloride ≥99.0% (by total nitrogen basis)

Proveedor: TCI
Atención

Sinónimos: 3-Amino-2-fluoropropionic Acid Hydrochloride

F0185-1G F0185-100MG
TCIAF0185-1GEA 419 EUR
TCIAF0185-1G TCIAF0185-100MG
2-Fluoro-β-alanine hydrochloride ≥99.0% (by total nitrogen basis)
2-Fluoro-beta-alanine hydrochloride
Formula: C₃H₆FNO₂
MW: 143.54 g/mol
Número MDL: MFCD00060202
Núm. CAS: 867-84-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 99.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR