3-Fluoro-4-n-octyloxybenzoic acid ≥97.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
F0337-1G
F0337-5G
TCIAF0337-1GEA
206
EUR
TCIAF0337-1G
TCIAF0337-5G
3-Fluoro-4-n-octyloxybenzoic acid ≥97.0% (by GC, titration analysis)
3-Fluoro-4-n-octyloxybenzoic acid
Formula:
C₁₅H₂₁FO₃ |
Número MDL:
MFCD00191649 Núm. CAS: 326-78-3 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...