4-(4-Fluorobenzoyl)butyric acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 5-(4-Fluorophenyl)-5-oxovaleric Acid, 5-(4-Fluorophenyl)-5-oxopentanoic acid

F0476-1G F0476-5G
TCIAF0476-1GEA 24 EUR
TCIAF0476-1G TCIAF0476-5G
4-(4-Fluorobenzoyl)butyric acid ≥98.0% (by GC, titration analysis)
4-(4-Fluorobenzoyl)butyric acid
Formula: C₁₁H₁₁FO₃
MW: 210.20 g/mol
Número MDL: MFCD00667222
Núm. CAS: 149437-76-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 141.0 to 145.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR