(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: N-Fmoc-L-aspartic Acid 1-tert-Butyl Ester, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 1-tert-Butyl Ester, Fmoc-Asp(OH)-OtBu, Fmoc-Asp-OtBu, 1-tert-Butyl N-Fmoc-L-aspartate

F0503-1G F0503-5G
TCIAF0503-1GEA 18 EUR
TCIAF0503-1G TCIAF0503-5G
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid ≥98.0% (by HPLC, titration analysis)
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid
Formula: C₂₃H₂₅NO₆
MW: 411,5 g/mol
Número MDL: MFCD00065631
Núm. CAS: 129460-09-9

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -20.0 to -28.0 deg (C=1, DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR