5-Fluoro-2-methoxy-4-pyrimidinone ≥98.0% (by GC, titration analysis)
Proveedor: TCI
Sinónimos:
5-Fluoro-2-methoxypyrimidin-4(3H)-one
F0547-25G
F0547-5G
TCIAF0547-25GEA
51
EUR
TCIAF0547-25G
TCIAF0547-5G
5-Fluoro-2-methoxy-4-pyrimidinone ≥98.0% (by GC, titration analysis)
5-Fluoro-2-methoxy-4-pyrimidinone
Formula:
C₅H₅FN₂O₂ MW: 144,11 g/mol |
Número MDL:
MFCD03265437 Núm. CAS: 1480-96-2 EINECS: 473-810-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 198.0 to 202.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...