4-Fluoropyridine hydrochloride ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
F0628-1G
F0628-5G
TCIAF0628-1GEA
94
EUR
TCIAF0628-1G
TCIAF0628-5G
4-Fluoropyridine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Fluoropyridine hydrochloride
Formula:
C₅H₅ClFN MW: 133.55 g/mol Pto. de fusión: 100°C |
Número MDL:
MFCD00234055 Núm. CAS: 39160-31-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...