Fleroxacin ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Megalone, 6,8-Difluoro-1-(2-fluoroethyl)-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic Acid, 6,8-Difluoro-1-(2-fluoroethyl)-7-(4-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
F0646-25G
F0646-5G
TCIAF0646-25GEA
311
EUR
TCIAF0646-25G
TCIAF0646-5G
Fleroxacin ≥98.0% (by titrimetric analysis)
Fleroxacin
Formula:
C₁₇H₁₈F₃N₃O₃ MW: 369,34 g/mol |
Número MDL:
MFCD00864880 Núm. CAS: 79660-72-3 Índice Merck: 14,04099 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...