N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
Fmoc-Cys(Trt)-OH, N-Fmoc-S-trityl-L-cysteine
F0652-25G
F0652-5G
TCIAF0652-25GEA
63
EUR
TCIAF0652-25G
TCIAF0652-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine ≥98.0% (by HPLC, titration analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine
Formula:
C₃₇H₃₁NO₄S MW: 585,72 g/mol |
Número MDL:
MFCD00038538 Núm. CAS: 103213-32-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 173.0 to 177.0 deg-C |
Specific rotation [a]20/D | +15.0 to +19.0 deg (C=1, THF) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...