N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-2-phenylglycine ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-phenylacetic acid, N-Fmoc-L-α-phenylglycine, Fmoc-Phg-OH, N-Fmoc-L-2-phenylglycine
F0669-1G
F0669-5G
TCIAF0669-1GEA
16
EUR
TCIAF0669-1G
TCIAF0669-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-2-phenylglycine ≥98.0% (by HPLC, titration analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-2-phenylglycine
Formula:
C₂₃H₁₉NO₄ MW: 373,41 g/mol |
Número MDL:
MFCD00155632 Núm. CAS: 102410-65-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 174.0 to 178.0 deg-C |
Specific rotation [a]20/D | +78.0 to +84.0 deg (C=1, DMF) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...