Fluconazole ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-(2,4-Difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol, 2-(2,4-Difluorophenyl)-1,3-di(1H-1,2,4-triazol-1-yl)propan-2-ol
F0677-1G
F0677-5G
TCIAF0677-1GEA
48
EUR
TCIAF0677-1G
TCIAF0677-5G
Fluconazole ≥98.0% (by GC, titration analysis)
Fluconazole
Formula:
C₁₃H₁₂F₂N₆O MW: 306,28 g/mol |
Número MDL:
MFCD00274549 Núm. CAS: 86386-73-4 Índice Merck: 14,04122 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 138.0 to 142.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...