Felodipine ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Atención
|
Sinónimos:
Ethyl Methyl 4-(2,3-Dichlorophenyl)-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate, 3-Ethyl 5-methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, 4-(2,3-Dichlorophenyl)-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic Acid Ethyl Methyl Ester
F0814-1G
F0814-5G
TCIAF0814-1GEA
91
EUR
TCIAF0814-1G
TCIAF0814-5G
Felodipine ≥98.0% (by HPLC, total nitrogen)
Felodipine
Formula:
C₁₈H₁₉Cl₂NO₄ MW: 384,25 g/mol |
Número MDL:
MFCD00868316 Núm. CAS: 72509-76-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 142.0 to 146.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...