Flubendazole ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
Methyl (5-(4-fluorobenzoyl)-1H-benzo[d]imidazol-2-yl)carbamate, Methyl [5-(4-Fluorobenzoyl)benzimidazol-2-yl]carbamate, [5-(4-Fluorobenzoyl)benzimidazol-2-yl]carbamic acid Methyl Ester
F0825-5G
F0825-25G
TCIAF0825-5GEA
45
EUR
TCIAF0825-5G
TCIAF0825-25G
Flubendazole ≥98.0% (by HPLC, titration analysis)
Flubendazole
Formula:
C₁₆H₁₂FN₃O₃ MW: 313,29 g/mol Pto. de fusión: 260°C |
Número MDL:
MFCD00871999 Núm. CAS: 31430-15-6 EINECS: 250-624-4 Índice Merck: 14,04118 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...