6-Fluoro-1H-indole-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 6-Fluoroindole-2-carboxylic acid

F0835-200MG F0835-1G
TCIAF0835-200MGEA 90 EUR
TCIAF0835-200MG TCIAF0835-1G
6-Fluoro-1H-indole-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)
6-Fluoro-1H-indole-2-carboxylic acid
Formula: C₉H₆FNO₂
MW: 179,15 g/mol
Número MDL: MFCD01863162
Núm. CAS: 3093-97-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05dc7d2547a6c46eb23841aa37ab94b91ab2076f&vid=05dc7d2547a6c46eb23841aa37ab94b91ab2076f&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR