Forskolin ≥95.0% (by HPLC)
Proveedor: TCI
Sinónimos:
(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate
F0855-10MG
F0855-50MG
TCIAF0855-10MGEA
87
EUR
TCIAF0855-10MG
TCIAF0855-50MG
Forskolin ≥95.0% (by HPLC)
Forskolin
Formula:
C₂₂H₃₄O₇ |
Número MDL:
MFCD00082317 Núm. CAS: 66575-29-9 EINECS: 266-410-9 Índice Merck: 14,02476 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Melting point | 224.0 to 228.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...