4-Fluoro-L-2-phenylglycine ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
4-Fluoro-L-phenylglycine, (S)-2-Amino-2-(4-fluorophenyl)acetic Acid, H-Phg(4-F)-OH, (S)-4-Fluorophenylglycine
F0862-5G
F0862-1G
TCIAF0862-5GEA
167
EUR
TCIAF0862-5G
TCIAF0862-1G
4-Fluoro-L-2-phenylglycine ≥98.0% (by HPLC, titration analysis)
4-Fluoro-L-2-phenylglycine
Formula:
C₈H₈FNO₂ MW: 169,16 g/mol |
Número MDL:
MFCD00213799 Núm. CAS: 19883-57-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +135 to +145 deg (C=1, 1mol/L HCl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...