N-[(9H-Fluoren-9-ylmethoxy)carbonyl]pentafluoro-L-phenylalanine ≥97.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
N-Fmoc-pentafluoro-L-phenylalanine, Fmoc-Phe(F5)-OH
F0900-200MG
F0900-1G
TCIAF0900-200MGEA
36
EUR
TCIAF0900-200MG
TCIAF0900-1G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]pentafluoro-L-phenylalanine ≥97.0% (by HPLC, titration analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]pentafluoro-L-phenylalanine
Formula:
C₂₄H₁₆F₅NO₄ MW: 477.39 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00270612 Núm. CAS: 205526-32-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Specific rotation [a]20/D | -18.0 to -21.0 deg (C=1,MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...