4-Fluoro-D-phenylalanine ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
(R)-2-Amino-3-(4-fluorophenyl)propanoic acid, H-D-Phe(4-F)-OH
F0901-1G
F0901-5G
TCIAF0901-1GEA
41
EUR
TCIAF0901-1G
TCIAF0901-5G
4-Fluoro-D-phenylalanine ≥98.0% (by HPLC, titration analysis)
4-Fluoro-D-phenylalanine
Formula:
C₉H₁₀FNO₂ |
Número MDL:
MFCD00063063 Núm. CAS: 18125-46-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +23.0 to +27.0 deg (C=1,H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...