Fmoc-4-fluoro-L-phenylalanine ≥95.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-fluoro-L-phenylalanine, Fmoc-p-fluoro-Phe-OH, Fmoc-Phe(4-F)-OH, (S)-2-[[[(9H-Fluoren-9-yl)methoxy]carbonyl]amino]-3-(4-fluorophenyl)propanoic acid, N-Fmoc-4-fluoro-L-phenylalanine
F0902-1G
F0902-5G
TCIAF0902-1GEA
61
EUR
TCIAF0902-1G
TCIAF0902-5G
Fmoc-4-fluoro-L-phenylalanine ≥95.0% (by HPLC, titration analysis)
Fmoc-4-fluoro-L-phenylalanine
Formula:
C₂₄H₂₀FNO₄ MW: 405.425 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00191197 Núm. CAS: 169243-86-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 182.0 to 186.0 deg-C |
Specific rotation [a]20/D | -36.0 to -40.0 deg (C=1,DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...