2-Fluoro-5-methylphenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-Fluoro-5-methylbenzeneboronic acid, 2-Fluoro-5-methylboronic acid
F0915-1G
F0915-5G
TCIAF0915-1GEA
23
EUR
TCIAF0915-1G
TCIAF0915-5G
2-Fluoro-5-methylphenylboronic acid (contains varying amounts of Anhydride)
2-Fluoro-5-methylphenylboronic acid
Formula:
C₇H₈BFO₂ MW: 153,9466 g/mol |
Número MDL:
MFCD03428512 Núm. CAS: 166328-16-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 113.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...