Fmoc-d-pipecolic acid ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
(R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-2-carboxylic acid, 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-pipecolic acid, (R)-1-Fmoc-2-piperidinecarboxylic acid, Fmoc-D-Pip-OH, 1-Fmoc-D-pipecolic acid, Fmoc-D-Homopro-OH, (R)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid
F0937-1G
F0937-5G
TCIAF0937-1GEA
35
EUR
TCIAF0937-1G
TCIAF0937-5G
Fmoc-d-pipecolic acid ≥98.0% (by HPLC, titration analysis)
Fmoc-d-pipecolic acid
Formula:
C₂₁H₂₁NO₄ MW: 351,402 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00235899 Núm. CAS: 101555-63-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 155.0 to 159.0 deg-C |
Specific rotation [a]20/D | +21.0 to +25.0 deg (C=1, MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...